3,5-Diisopropylpyrazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3,5-Diisopropyl-1H-pyrazole
OR921043-1G
OR921043-25G
OR921043-5G
APOSOR921043-1GEA
458
DKK
APOSOR921043-1G
APOSOR921043-25G
APOSOR921043-5G
3,5-Diisopropylpyrazole
3,5-Diisopropylpyrazole
Formel:
C₉H₁₆N₂ Smeltepunkt: 88 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00671516 CAS-nummer: 17536-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...