8-Methoxyquinaldine 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR920706-1G
OR920706-5G
APOSOR920706-1GEA
305
DKK
APOSOR920706-1G
APOSOR920706-5G
8-Methoxyquinaldine 95%
8-Methoxyquinaldine
Formel:
C₁₁H₁₁NO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00034436 CAS-nummer: 3033-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...