3,5-Dimethylcyclohexanol (mixture of isomers) 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3,5-Dimethylcyclohexanol

OR920486-100G OR920486-25G OR920486-5G
APOSOR920486-100GEA 4670 DKK
APOSOR920486-100G APOSOR920486-25G APOSOR920486-5G
3,5-Dimethylcyclohexanol (mixture of isomers) 95%
3,5-Dimethylcyclohexanol (mixture of isomers)
Formel: C₈H₁₆O
Molekylvægt: 128.2120 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001447
CAS-nummer: 5441-52-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR