1-(2-Chlorethoxy)propan 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Chloroethyln-propylether, 2-Chloroethyl Propyl ether, 2-Propoxyethyl chloride

OR919962-25G OR919962-100G
APOSOR919962-25GEA 545 DKK
APOSOR919962-25G APOSOR919962-100G
1-(2-Chlorethoxy)propan 98%
1-(2-Chloroethoxy)propane
Formel: ClCH₂CH₂OCH₂CH₂CH₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00059052
CAS-nummer: 42149-74-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR