O-Allylhydroxylamine hydrochloride 95%
Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
OR916474-25G OR916474-5G
APOSOR916474-25GEA 441 DKK
APOSOR916474-25G APOSOR916474-5G
O-Allylhydroxylamine hydrochloride 95%
O-Allylhydroxylamine hydrochloride
Formel: C₃H₇NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012957
CAS-nummer: 38945-21-0