(2-Hydroxyethyl)hydrazin 92%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
β-Hydroxyethylhydrazine
OR9124-100G
OR9124-10G
OR9124-500G
OR9124-250G
APOSOR9124-100GEA
458
DKK
APOSOR9124-100G
APOSOR9124-10G
APOSOR9124-500G
APOSOR9124-250G
(2-Hydroxyethyl)hydrazin 92%
(2-Hydroxyethyl)hydrazine
Formel:
C₂H₈N₂O Kogepunkt: 218…220 °C Smeltepunkt: –70 °C Massefylde: 1,123 g/cm³ (25 °C) Flammepunkt: 77 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007623 CAS-nummer: 109-84-2 EINECS: 203-711-6 UN nummer: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...