Pirfenidone 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-Methyl-1-phenylpyridin-2(1H)-one, 5-Methyl-1-phenyl-2-pyridone

OR911941-10G OR911941-1G OR911941-5G
APOSOR911941-10GEA 830 DKK
APOSOR911941-10G APOSOR911941-1G APOSOR911941-5G
Pirfenidone 98%
Pirfenidone
Formel: C₁₂H₁₁NO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00866047
CAS-nummer: 53179-13-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR