4'-Ethylacetophenon 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR911728-100G OR911728-500G
APOSOR911728-100GEA 291 DKK
APOSOR911728-100G APOSOR911728-500G
4'-Ethylacetophenon 98%
4'-Ethylacetophenon
Formel: C₁₀H₁₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009262
CAS-nummer: 937-30-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR