1-Bromo-4-methoxybutane 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR911726-100G OR911726-25G
APOSOR911726-100GEA 2180 DKK
APOSOR911726-100G APOSOR911726-25G
1-Bromo-4-methoxybutane 98%
1-Bromo-4-methoxybutane
Formel: C₅H₁₁BrO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00037102
CAS-nummer: 4457-67-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR