5-Nitroindazol 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Nitro-1H-indazole, 5-NINA, 5-Nitroindazole

OR9104-100G OR9104-500G OR9104-25G
APOSOR9104-100GEA 523 DKK
APOSOR9104-100G APOSOR9104-500G APOSOR9104-25G
5-Nitroindazol 95%
5-Nitroindazol
Formel: C₇H₅N₃O₂
Smeltepunkt: 204…208 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005693
CAS-nummer: 5401-94-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR