3-(4-Chlorophenyl)-1,2,4-oxadiazole 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR910107-100G
OR910107-25G
OR910107-5G
APOSOR910107-100GEA
6050
DKK
APOSOR910107-100G
APOSOR910107-25G
APOSOR910107-5G
3-(4-Chlorophenyl)-1,2,4-oxadiazole 98%
3-(4-Chlorophenyl)-1,2,4-oxadiazole
Formel:
C₈H₅ClN₂O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00121590 CAS-nummer: 10550-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...