4-Chloro-8-methylquinazoline 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR909777-1G OR909777-5G
APOSOR909777-1GEA 709 DKK
APOSOR909777-1G APOSOR909777-5G
4-Chloro-8-methylquinazoline 97%
4-Chloro-8-methylquinazoline
Formel: C₉H₇ClN₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08457971
CAS-nummer: 58421-80-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR