1-Boc-4-cyano-4-methylpiperidine 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: tert-Butyl 4-cyano-4-methylpiperidine-1-carboxylate

OR907558-1G OR907558-250MG OR907558-500MG
APOSOR907558-1GEA 4320 DKK
APOSOR907558-1G APOSOR907558-250MG APOSOR907558-500MG
1-Boc-4-cyano-4-methylpiperidine 97%
1-Boc-4-cyano-4-methylpiperidine
Formel: C₁₂H₂₀N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11849789
CAS-nummer: 530115-96-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR