1-Methyl-1H-indole-2-carbaldehyde 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR906982-1G
OR906982-5G
APOSOR906982-1GEA
491
DKK
APOSOR906982-1G
APOSOR906982-5G
1-Methyl-1H-indole-2-carbaldehyde 98%
1-Methyl-1H-indole-2-carbaldehyde
Formel:
C₁₀H₉NO Smeltepunkt: 81…85 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01863665 CAS-nummer: 27421-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...