Methyl-4-amino-2-chloropyrimidine-5-carboxylate 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methyl 4-amino-2-chloropyrimidine-5-carboxylate

OR906639-1G OR906639-250MG
APOSOR906639-1GEA 5780 DKK
APOSOR906639-1G APOSOR906639-250MG
Methyl-4-amino-2-chloropyrimidine-5-carboxylate 97%
Methyl-4-amino-2-chloropyrimidine-5-carboxylate
Formel: C₆H₆ClN₃O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14585039
CAS-nummer: 858269-13-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR