5-Bromo-6-methylpyrimidin-4-amine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR906006-100MG OR906006-1G OR906006-250MG
APOSOR906006-100MGEA 0 DKK
APOSOR906006-100MG APOSOR906006-1G APOSOR906006-250MG
5-Bromo-6-methylpyrimidin-4-amine 95%
5-Bromo-6-methylpyrimidin-4-amine
Formel: C₅H₆BrN₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00626351
CAS-nummer: 7752-48-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR