4-Chloro-3-hydroxybenzaldehyde 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR904781-10G OR904781-1G OR904781-5G
APOSOR904781-10GEA 1940 DKK
APOSOR904781-10G APOSOR904781-1G APOSOR904781-5G
4-Chloro-3-hydroxybenzaldehyde 98%
4-Chloro-3-hydroxybenzaldehyde
Formel: C₇H₅ClO₂
Molekylvægt: 156.5700 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16999770
CAS-nummer: 56962-12-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR