4-(Thiazol-2-yl)aniline 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR904473-1G OR904473-250MG
APOSOR904473-1GEA 4580 DKK
APOSOR904473-1G APOSOR904473-250MG
4-(Thiazol-2-yl)aniline 95%
4-(Thiazol-2-yl)aniline
Formel: C₉H₈N₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06655097
CAS-nummer: 193017-26-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR