3-Phenylpyridin-2-amine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR903930-1G OR903930-250MG
APOSOR903930-1GEA 6180 DKK
APOSOR903930-1G APOSOR903930-250MG
3-Phenylpyridin-2-amine 95%
3-Phenylpyridin-2-amine
Formel: C11H10N2
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04114119
CAS-nummer: 87109-10-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR