6-Bromo[1,2,4]triazolo[1,5-a]pyrazine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR903771-1G OR903771-250MG
APOSOR903771-1GEA 366 DKK
APOSOR903771-1G APOSOR903771-250MG
6-Bromo[1,2,4]triazolo[1,5-a]pyrazine 95%
6-Bromo[1,2,4]triazolo[1,5-a]pyrazine
Formel: C₅H₃BrN₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12923113
CAS-nummer: 1233026-51-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR