2-Chloro-5-isopropylpyrimidine 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR903243-1G OR903243-250MG
APOSOR903243-1GEA 3820 DKK
APOSOR903243-1G APOSOR903243-250MG
2-Chloro-5-isopropylpyrimidine 95%
2-Chloro-5-isopropylpyrimidine
Formel: C₇H₉ClN₂
Molekylvægt: 156.6100 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11109833
CAS-nummer: 596114-50-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR