4-Iodoisoxazole 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR902256-1G OR902256-5G
APOSOR902256-1GEA 284 DKK
APOSOR902256-1G APOSOR902256-5G
4-Iodoisoxazole 97%
4-Iodoisoxazole
Formel: C₃H₂INO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09999222
CAS-nummer: 847490-69-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR