2-(Cyclopropylamino)-3-nitro-5-picoline 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Cyclopropylamino-5-methyl-3-nitropyridine

OR902003-1G OR902003-25G OR902003-5G
APOSOR902003-1GEA 985 DKK
APOSOR902003-1G APOSOR902003-25G APOSOR902003-5G
2-(Cyclopropylamino)-3-nitro-5-picoline 98%
2-(Cyclopropylamino)-3-nitro-5-picoline
Formel: C₉H₁₁N₃O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10699715
CAS-nummer: 1033202-65-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR