3-(BOC-amino)pyridin 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR901723-1G OR901723-5G
APOSOR901723-1GEA 0 DKK
APOSOR901723-1G APOSOR901723-5G
3-(BOC-amino)pyridin 95%
3-(BOC-amino)pyridin
Formel: C₁₀H₁₄N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02179231
CAS-nummer: 56700-70-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR