8-Methyl-2-quinolinol 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 8-Methylquinolin-2(1H)-one, 8-Methyl-2(1H)-quinolinone

OR901652-1G OR901652-5G
APOSOR901652-1GEA 371 DKK
APOSOR901652-1G APOSOR901652-5G
8-Methyl-2-quinolinol 98%
8-Methyl-2-quinolinol
Formel: C₁₀H₉NO
Molekylvægt: 159.1846 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13195790
CAS-nummer: 4053-36-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR