2-(Chlormethyl)benzimidazol 96%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-(Chloromethyl)-1H-benzimidazole
OR9012-5G
OR9012-25G
OR9012-100G
OR9012-1G
OR9012-10G
APOSOR9012-5GEA
218
DKK
APOSOR9012-5G
APOSOR9012-25G
APOSOR9012-100G
APOSOR9012-1G
APOSOR9012-10G
2-(Chlormethyl)benzimidazol 96%
2-(Chloromethyl)benzimidazole
Formel:
C₈H₇ClN₂ Smeltepunkt: 141…148 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005602 CAS-nummer: 4857-04-9 EINECS: 225-452-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...