4-(Diethylamino)benzonitril 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR900360-100G OR900360-25G OR900360-5G
APOSOR900360-100GEA 4860 DKK
APOSOR900360-100G APOSOR900360-25G APOSOR900360-5G
4-(Diethylamino)benzonitril 98%
4-(Diethylamino)benzonitril
Formel: C₁₁H₁₄N₂
Molekylvægt: 174.25 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00019754
CAS-nummer: 2873-90-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR