2-Chlor-6-methylpyridin
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR8825-100G
OR8825-25G
OR8825-5G
APOSOR8825-100GEA
632
DKK
APOSOR8825-100G
APOSOR8825-25G
APOSOR8825-5G
2-Chlor-6-methylpyridin
2-Chloro-6-methylpyridine
Formel:
C₆H₆ClN Kogepunkt: 64…68 °C/10mmHg Massefylde: 1,167 g/cm³ (25 °C) Flammepunkt: 74 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006245 CAS-nummer: 18368-63-3 EINECS: 242-241-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...