(R)-(+)-4-Benzyl-2-oxazolidinon
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(R)-4-Benzyl-2-oxazolidinone
OR8727-5G
OR8727-1G
APOSOR8727-5GEA
164
DKK
APOSOR8727-5G
APOSOR8727-1G
(R)-(+)-4-Benzyl-2-oxazolidinon
(R)-(+)-4-Benzyl-2-oxazolidinon
Formel:
C₁₀H₁₁NO₂ Smeltepunkt: 86…90 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010846 CAS-nummer: 102029-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...