(-)-Dibenzoyl-L-tartaric acid monohydrate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate

OR8726-1KG OR8726-250G OR8726-5KG
APOSOR8726-1KGEA 491 DKK
APOSOR8726-1KG APOSOR8726-250G APOSOR8726-5KG
(-)-Dibenzoyl-L-tartaric acid monohydrate
(-)-Dibenzoyl-L-tartaric acid monohydrate

Optical resolving agent

Formel: C₁₈H₁₄O₈·1H₂O
Smeltepunkt: 88…89 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149119
CAS-nummer: 62708-56-9
EINECS: 613-072-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR