2-Amino-5-chlor-3-nitropyridin
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Chloro-3-nitro-2-pyridylamine, 5-Chloro-3-nitropyridin-2-amine
OR8629-25G
OR8629-100G
APOSOR8629-25GEA
450
DKK
APOSOR8629-25G
APOSOR8629-100G
2-Amino-5-chlor-3-nitropyridin
2-Amino-5-chloro-3-nitropyridine
Formel:
C₅H₄ClN₃O₂ Smeltepunkt: 193…197 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00092011 CAS-nummer: 5409-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...