4-Aminopyrimidin 97+%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR8287-25G
OR8287-5G
OR8287-1G
APOSOR8287-25GEA
1370
DKK
APOSOR8287-25G
APOSOR8287-5G
APOSOR8287-1G
4-Aminopyrimidin 97+%
4-Aminopyrimidine
Formel:
C₄H₅N₃ Smeltepunkt: 147…156 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006112 CAS-nummer: 591-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...