2-Nitroresorcinol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Nitrobenzene-1,3-diol

OR8239-100G OR8239-25G OR8239-500G
APOSOR8239-100GEA 1180 DKK
APOSOR8239-100G APOSOR8239-25G APOSOR8239-500G
2-Nitroresorcinol
2-Nitroresorcinol
Formel: C₆H₅NO₄
Smeltepunkt: 81…83 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007124
CAS-nummer: 601-89-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR