1-Bromo-6-phenylhexane 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(6-Bromohex-1-yl)benzene
OR8181-1G
OR8181-5G
APOSOR8181-1GEA
230
DKK
APOSOR8181-1G
APOSOR8181-5G
1-Bromo-6-phenylhexane 98%
1-Bromo-6-phenylhexane
Formel:
C₆H₅(CH₂)₆Br Molekylvægt: 241.18 g/mol Kogepunkt: 77…78 °C/0.075mmHg Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00236022 CAS-nummer: 27976-27-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...