2-Chlor-4-pyridinboronsyrepinacolester 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Chloropyridine-4-boronic acid pinacol ester, 2-(2-Chloro-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloropyridine-4-boronic acid Pinacol ester

OR7616-25G OR7616-5G
APOSOR7616-25GEA 2040 DKK
APOSOR7616-25G APOSOR7616-5G
2-Chlor-4-pyridinboronsyrepinacolester 97%
2-Chlor-4-pyridinboronsyrepinacolester
Formel: C₁₁H₁₅BClNO₂
Molekylvægt: 239.51 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04039870
CAS-nummer: 458532-84-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR