Diisopropyl(bromomethyl)boronate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (Bromomethyl)boronic Acid Diisopropyl Ester

OR7201-1G OR7201-5G
APOSOR7201-1GEA 1430 DKK
APOSOR7201-1G APOSOR7201-5G
Diisopropyl(bromomethyl)boronate 95%
Diisopropyl(bromomethyl)boronate
Formel: C₇H₁₆BBrO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01631222
CAS-nummer: 137297-49-5
UN nummer: 3272
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR