4-Cyclopentylphenylboronic acid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR7191-1G OR7191-250MG
APOSOR7191-1GEA 2060 DKK
APOSOR7191-1G APOSOR7191-250MG
4-Cyclopentylphenylboronic acid 98%
4-Cyclopentylphenylboronic acid
Formel: C₁₁H₁₅BO₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04039014
CAS-nummer: 945837-57-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR