Benzo[b]thiophene-2-carboxylic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR7075-100G OR7075-25G
APOSOR7075-100GEA 1910 DKK
APOSOR7075-100G APOSOR7075-25G
Benzo[b]thiophene-2-carboxylic acid 97%
Benzo[b]thiophene-2-carboxylic acid
Formel: C₉H₆O₂S
Smeltepunkt: 238…240 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00051636
CAS-nummer: 6314-28-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR