tert-Butyl-3-oxopropylcarbamate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR70078-1G OR70078-250MG
APOSOR70078-1GEA 2460 DKK
APOSOR70078-1G APOSOR70078-250MG
tert-Butyl-3-oxopropylcarbamate
tert-Butyl-3-oxopropylcarbamate
Formel: C₈H₁₅NO₃
Opbevaringstemperatur: Fryser
MDL Number: MFCD08059952
CAS-nummer: 58885-60-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR