1-(Cyclohexylmethyl)piperazine 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR6904-1G OR6904-500MG
APOSOR6904-1GEA 382 DKK
APOSOR6904-1G APOSOR6904-500MG
1-(Cyclohexylmethyl)piperazine 97%
1-(Cyclohexylmethyl)piperazine
Formel: C₁₁H₂₂N₂
Kogepunkt: 230…231 °C
Massefylde: 0,938 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01075237
CAS-nummer: 57184-23-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR