3-Nitro-o-phenylendiamin 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Nitrobenzene-1,2-diamine, 3-Nitro-1,2-phenylenediamine

OR6780-1G OR6780-5G OR6780-10G OR6780-25G
APOSOR6780-1GEA 208 DKK
APOSOR6780-1G APOSOR6780-5G APOSOR6780-10G APOSOR6780-25G
3-Nitro-o-phenylendiamin 97%
3-Nitro-o-phenylendiamin
Formel: C₆H₇N₃O₂
Molekylvægt: 153.14 g/mol
Smeltepunkt: 156…162 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007722
CAS-nummer: 3694-52-8
EINECS: 223-013-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR