4,4'-Diiodbiphenyl 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4,4'-Diiodo-1,1'-biphenyl
OR6696-100G
OR6696-25G
APOSOR6696-100GEA
1620
DKK
APOSOR6696-100G
APOSOR6696-25G
4,4'-Diiodbiphenyl 98%
4,4'-Diiodbiphenyl
Formel:
C₁₂H₈I₂ Smeltepunkt: 201…204 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001057 CAS-nummer: 3001-15-8 EINECS: 221-080-5 UN nummer: 3152 ADR: 9,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...