Cycloheptancarboxylsyre 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR6532-5G OR6532-1G
APOSOR6532-5GEA 393 DKK
APOSOR6532-5G APOSOR6532-1G
Cycloheptancarboxylsyre 95%
Cycloheptancarboxylsyre
Formel: C₈H₁₄O₂
Kogepunkt: 135…138 °C/19mmHg
Massefylde: 1,035 g/cm³ (25 °C)
Flammepunkt: >113 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004152
CAS-nummer: 1460-16-8
EINECS: 215-954-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR