4-Dibenzofuranboronic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR6444-1G OR6444-5G OR6444-25G
APOSOR6444-1GEA 360 DKK
APOSOR6444-1G APOSOR6444-5G APOSOR6444-25G
4-Dibenzofuranboronic acid
4-Dibenzofuranboronic acid
Formel: C₁₂H₉BO₃
Smeltepunkt: 286…291 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00092336
CAS-nummer: 100124-06-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR