3-(2-Aminoethyl)thiazolidine-2,4-dione hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR6437-1G OR6437-5G OR6437-500MG
APOSOR6437-1GEA 414 DKK
APOSOR6437-1G APOSOR6437-5G APOSOR6437-500MG
3-(2-Aminoethyl)thiazolidine-2,4-dione hydrochloride
3-(2-Aminoethyl)thiazolidine-2,4-dione hydrochloride
Formel: C₅H₉ClN₂O₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00801011
CAS-nummer: 19382-49-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR