Ethyl phenylglyoxylate 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR6395-100G OR6395-50G
APOSOR6395-100GEA 1660 DKK
APOSOR6395-100G APOSOR6395-50G
Ethyl phenylglyoxylate 98%
Ethyl phenylglyoxylate
Formel: C₁₀H₁₀O₃
Kogepunkt: 138…139 °C
Massefylde: 1,122 g/cm³ (25 °C)
Flammepunkt: 113 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009120
CAS-nummer: 1603-79-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR