Benzofuran-2-carboxylic acid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR6305-100G OR6305-25G OR6305-5G
APOSOR6305-100GEA 4310 DKK
APOSOR6305-100G APOSOR6305-25G APOSOR6305-5G
Benzofuran-2-carboxylic acid 98%
Benzofuran-2-carboxylic acid
Formel: C₉H₆O₃
Smeltepunkt: 193…196 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005848
CAS-nummer: 496-41-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR