2,1,3-Benzothiadiazol-4-ylamine 95+%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR6147-1G OR6147-5G
APOSOR6147-1GEA 327 DKK
APOSOR6147-1G APOSOR6147-5G
2,1,3-Benzothiadiazol-4-ylamine 95+%
2,1,3-Benzothiadiazol-4-ylamine
Formel: C₆H₅N₃S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005810
CAS-nummer: 767-64-6
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR