4-Amino-3-chlorbenzoesyre 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR61423-1G OR61423-25G OR61423-5G OR61423-100G
APOSOR61423-1GEA 110 DKK
APOSOR61423-1G APOSOR61423-25G APOSOR61423-5G APOSOR61423-100G
4-Amino-3-chlorbenzoesyre 97%
4-Amino-3-chlorbenzoesyre
Formel: C₇H₆ClNO₂
Smeltepunkt: 224…229 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00460423
CAS-nummer: 2486-71-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR