1-Nitro-2,3,4,5-tetrachlorbenzen
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2,3,4,5-Tetrachloronitrobenzene
OR61402-25G
OR61402-5G
APOSOR61402-25GEA
13680
DKK
APOSOR61402-25G
APOSOR61402-5G
1-Nitro-2,3,4,5-tetrachlorbenzen
1-Nitro-2,3,4,5-tetrachlorbenzen
Formel:
C₆HCl₄NO₂ Smeltepunkt: 65…67 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007064 CAS-nummer: 879-39-0 EINECS: 212-906-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...